Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, MP Biomedicals™

Catalog No. MP215775005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215775005 5 g
MP215775025 25 g
MP215775080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215775005 Supplier MP Biomedicals, Inc Supplier No. 0215775005
Add to Cart
Add to Cart

5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, White Powder

  • It is useful as a nonionic density gradient medium.
  • It is a Non-ionic monomeric triiodinated x-ray contrast agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66108-95-0
Molecular Formula C19H26I3N3O9
Molecular Weight (g/mol) 821.14
MDL Number MFCD00077732
InChI Key NTHXOOBQLCIOLC-UHFFFAOYNA-N
Synonym Nycodenz, Histodenz™

Specifications

Color White
Quantity 5 g
Beilstein 2406632
Physical Form Powder
Assay 0.99
RTECSNumber CZ2205000
storageNote1 Store at -0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.