Learn More
Cayman Chemical 5-(N-ethyl-N-isopropyl)-Amiloride, 1154-25-2, 10 mg
SDP
Supplier:  Cayman Chemical 1440610
Encompass_Preferred
Molecular Formula C11H18ClN7O, Purity ≥98%, Formula Weight 299.8, Synonyms: 3-amino-N-(aminoiminomethyl)-6-chloro-5-[ethyl(1-methylethyl)amino]-2-pyrazinecarboxamide, EIPA, L-593,754, MH 12-43; A potent inhibitor of several NHE isoforms, inhibiting NHE1, NHE2, NHE3, and NHE5 with Ki values of 0.02, 0.5, 2.4, and 0.42 M, respectively; less effectively inhibits NHE4 (IC50 10 M).
Catalog No. NC1107618
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.