Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitro-2-pyridinecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. N09301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09301G 1 g
N09305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09301G Supplier TCI America Supplier No. N09301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30651-24-2
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD04114182
InChI Key QKYRCTVBMNXTBT-UHFFFAOYSA-N
Synonym 5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid
PubChem CID 520488
IUPAC Name 5-nitropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 1 g
Formula Weight 168.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitro-2-pyridinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.