Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroisatin 98.0+%, TCI America™
SDP

Catalog No. N057225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N057225G 25 g
N05725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N057225G Supplier TCI America Supplier No. N057225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 611-09-6
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00005720
InChI Key UNMYHYODJHKLOC-UHFFFAOYSA-N
Synonym 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
PubChem CID 4669250
IUPAC Name 5-nitro-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2

Specifications

Melting Point 255°C
Quantity 25 g
Formula Weight 192.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroisatin
RTECSNumber NL7970000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.