Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitrosalicylic acid, 98%

Catalog No. AAA1301014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1301014 25 g
AAA1301006 5 g
AAA1301022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1301014 Supplier Thermo Scientific Chemicals Supplier No. A1301014
Add to Cart

CAS: 96-97-9 | C7H5NO5 | 183.119 g/mol

5-Nitrosalicylic acid is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrosalicylic acid is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic, pharmaceutical intermediate.

Solubility
Soluble in water 1g/1475ml .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-97-9
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00007338
InChI Key PPDRLQLKHRZIJC-UHFFFAOYSA-N
Synonym 5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid
PubChem CID 7318
ChEBI CHEBI:61281
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O

Specifications

Melting Point 228°C to 236°C
Density 1.65
Quantity 25 g
Beilstein 2213719
Merck Index 14,6632
Solubility Information Soluble in water 1g/1475ml .
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 5-Nitrosalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-548-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.