Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nonanone 98.0+%, TCI America™
SDP

Catalog No. N0294100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0294100ML 100 mL
N029425ML 25 mL
N0294500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0294100ML Supplier TCI America Supplier No. N0294100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 502-56-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009484
InChI Key WSGCRAOTEDLMFQ-UHFFFAOYSA-N
Synonym 5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
PubChem CID 10405
IUPAC Name nonan-5-one
SMILES CCCCC(=O)CCCC

Specifications

Color Yellow
Boiling Point 186°C
Quantity 100 mL
UN Number 1224
Formula Weight 142.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Nonanone
EINECSNumber (2)-0572&(2)-2475
RTECSNumber RA8230000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.