Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Norbornen-2-methanol (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. N097925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N097925G 25 g
N09795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N097925G Supplier TCI America Supplier No. N097925G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95-12-5
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00167571
InChI Key LUMNWCHHXDUKFI-UHFFFAOYSA-N
Synonym 5-norbornene-2-methanol, bicyclo 2.2.1 hept-5-en-2-ylmethanol, bicyclo 2.2.1 hept-5-ene-2-methanol, cyclol, 2-hydroxymethyl-5-norbornene, 5-hydroxymethyl-2-norbornene, 5-norbornene 2-methanol, 5-hydroxymethylbicyclo 2.2.1 hept-2-ene, 2-hydroxymethyl bicyclo 2.2.1 hept-5-ene
PubChem CID 78946
IUPAC Name 5-bicyclo[2.2.1]hept-2-enylmethanol
SMILES C1C2CC(C1C=C2)CO

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 124.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Norbornen-2-methanol (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.