Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-O-(4,4'-Dimethoxytrityl)thymidine 98.0+%, TCI America™
SDP

Catalog No. D356625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D356625G 25 g
D35665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D356625G Supplier TCI America Supplier No. D356625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40615-39-2
Molecular Formula C31H32N2O7
Molecular Weight (g/mol) 544.60
MDL Number MFCD00010113
InChI Key UBTJZUKVKGZHAD-QZGLRKMJNA-N
Synonym 5'-o-4,4'-dimethoxytrityl thymidine, dmt-t, thymidine, 5'-o-bis 4-methoxyphenyl phenylmethyl, 5'-o-4,4'-dimethoxytrityl thymidine dmt-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenylmethoxy methyl-4-hydroxyoxolan-2-yl-5-methylpyrimidine-2,4-dione, 5'-o-dimethoxytrityl-deoxythymidine, dmtr-dthd, dimethoxytrityl-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenyl-methoxy methyl-4-hydroxy-tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione
PubChem CID 162419
IUPAC Name 1-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C)C(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Melting Point 151°C
Color White
Quantity 25 g
Formula Weight 544.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5′-O-(4,4′-Dimethoxytrityl)thymidine
EINECSNumber (5)-6792
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.