Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Oxoazelaic Acid 96.0+%, TCI America™
SDP

Catalog No. O02581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02581G 1 g
O02585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02581G Supplier TCI America Supplier No. O02581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57822-06-7
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.21
MDL Number MFCD00075542
InChI Key GTCHZEFRDKAINX-UHFFFAOYSA-N
Synonym 5-Ketoazelaic Acid, 5-Oxononanedioic Acid
PubChem CID 269945
IUPAC Name 5-oxononanedioic acid
SMILES OC(=O)CCCC(=O)CCCC(O)=O

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 1 g
Formula Weight 202.21
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Oxoazelaic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.