Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5 Percent Palladium on Calcium Carbonate Powder, Unreduced, Dry, 3 microns, BASF Catalyst, Spectrum™ Chemical
SDP

Catalog No. 18607947 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-607-947 100 g
18-607-945 5 g
18-607-946 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-947 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SP109100G
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7440-05-3 | Pd | 106.42 g/mol

Chemical Identifiers

CAS 7440-05-3 , 471-34-1
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
IUPAC Name palladium
SMILES [Pd]

Specifications

Physical Form Powder or Liquid
Quantity 100 g
Grade Not Available
Packaging Amber Glass Bottle
Chemical Name or Material 5% Palladium on Calcium Carbonate Powder, Unreduced, Dry, 3 μ, BASF Catalyst

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.