Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenyl-2-furaldehyde, 96%

Catalog No. AC437310010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437310010 1 g Glass bottle
AC437310050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437310010 Supplier Thermo Scientific Chemicals Supplier No. 437310010
Add to Cart

CAS: 13803-39-9 | C11H8O2 | 172.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13803-39-9
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.19
InChI Key BMJHNNPEPBZULA-UHFFFAOYSA-N
Synonym 5-phenyl-2-furaldehyde, 2-formyl-5-phenylfuran, 5-phenyl-furan-2-carbaldehyde, 5-phenylfurfural, 2-furancarboxaldehyde,5-phenyl, 5-phenylfuraldehyde, acmc-209y7p, 5-phenyl-2-furancarboxaldehyde
PubChem CID 2769420
IUPAC Name 5-phenylfuran-2-carbaldehyde
SMILES C1=CC=C(C=C1)C2=CC=C(O2)C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 29°C to 33°C
Color Brown to Yellow
Boiling Point 104°C to 108°C (6.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.19
Percent Purity 96%
Physical Form Liquid or Solid
Chemical Name or Material 5-Phenyl-2-furaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.