Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenyltetrazole 99.0+%, TCI America™
SDP

Catalog No. P1109100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1109100G 100 g
P110925G 25 g
P1109500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P1109100G Supplier TCI America Supplier No. P1109100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18039-42-4
Molecular Formula C7H6N4
Molecular Weight (g/mol) 146.153
MDL Number MFCD00022388
InChI Key MARUHZGHZWCEQU-UHFFFAOYSA-N
Synonym 5-phenyl-1h-tetrazole, 5-phenyltetrazole, 1h-tetrazole, 5-phenyl, 2h-tetrazole, 5-phenyl, phenyltetrazole, 5-phenyl tetrazole, expandex 5pt, expandex ox 5pt, kempore 50xpt, 5-phenyltetrazole van
PubChem CID 87425
IUPAC Name 5-phenyl-2H-tetrazole
SMILES C1=CC=C(C=C1)C2=NNN=N2

Specifications

Melting Point 215°C
Color White
Quantity 100 g
UN Number 1325
Formula Weight 146.15
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenyltetrazole
EINECSNumber (5)-6042
RTECSNumber XF7538750
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.