Learn More
AdipoGen 5-ROX
SDP
Supplier:  AdipoGen CDXC0004M200
Encompass_Preferred
Chemical. CAS 216699-35-3. Formula C33H30N2O5. MW 534.6. Synthetic. 5-ROX is the purified isomer of 56-ROX. It is preferred for some complicated biological applications where reproducibility is really critical. The minor positional difference between 5-ROX and 6-ROX might significantly affect some biological properties of the underlying conjugates. Used for oligonucleotide labeling and automated DNA sequencing applications.
Catalog No. 50-256-1565
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.