Learn More
Cayman Chemical Lipoxin A4, 89663-86-5, 25 g
SDP
Supplier:  Cayman Chemical 9041025
Encompass_Preferred
Molecular Formula C20H32O5, Purity ≥95%, Formula Weight 352.5, Synonyms: LXA4, 5(S),6(R),15(S)-TriHETE; A trihydroxy fatty acid containing a conjugated tetraene produced by the metabolism of ()15-HETE or 15-HpETE with human leukocytes; promotes leukocyte activation, chemotaxis effects, natural killer cell inhibition, and monocyte migration and adhesion.
Catalog No. NC9019975
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.