Learn More
Cayman Chemical Zona Radiata salmon Monoclon
SDP
Supplier:  Cayman Chemical 100017500 UL
Encompass_Preferred
Antigen purified eggshell proteins (zona radiata proteins and small amounts of zona pellucida proteins) from Atlantic salmon (S. salar) Host mouse, clone MN-8C4 Cross-reactivity ( ) Atlantic salmon (S. salar) (reacts primarily with a and β monomers), rainbow trout (O. mykiss), Arctic char (S. alpinus), turbot (S. maximus), and cod (G. morhua) Zrp Application(s) WB and ELISA Isotype IgG1-κ
Catalog No. 50-288-8884
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.