Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Sulfoisophthalic acid sodium salt

Catalog No. 142794100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-279-4100G 100 g
14-279-4500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-279-4100G Supplier Sigma Aldrich Supplier No. 142794100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5-Sulfoisophthalic acid sodium salt is a sulfonate carboxylate ligand which can be used in a variety of applications such as catalysis, gas storage, magnetic materials, and luminescence.

Specifications

CAS 6362-79-4
Molecular Formula C8H5NaO7S
Linear Formula NaO3SC6H3-1,3-(CO2H)2
Quantity 100 g
MDL Number MFCD00007495
Synonym 5-Sodiosulfoisophthalic acid; 5-Sulfoisophthalic acid monosodium salt; Monosodium 5-sulfoisophthalate; Sodium 3,5-dicarboxybenzene-1-sulfonate; Sodium 5-sulfoisophthalate
Molecular Weight (g/mol) 268.18
Percent Purity 95%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.