Learn More
Bachem 5-TAMRA-Amyloid Beta Protein (1-42) Trifluoroacetate
SDP
Supplier:  Bachem 4090153.0100
Encompass_Preferred
5-TAMRA-Amyloid Beta Protein (1-42) Trifluoroacetate, 5-TAMRA-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 1802087-80-4, C228H331N57O64S, Mr 4926.55, 0.1mg. 5-TAMRA-Ab42.
Catalog No. 50-194-6622
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.