Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(tert-Butoxycarbonylamino)-1-pentanol 97.0+%, TCI America™
SDP

Catalog No. B286925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B286925G 25 g
B28695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B286925G Supplier TCI America Supplier No. B286925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75178-90-4
Molecular Formula C10H21NO3
Molecular Weight (g/mol) 203.28
MDL Number MFCD01862955
InChI Key DDGNGFVNTZJMMZ-UHFFFAOYSA-N
Synonym 5-boc-amino-1-pentanol, tert-butyl n-5-hydroxypentyl carbamate, tert-butyl 5-hydroxypentyl carbamate, 5-tert-butoxycarbonylamino-1-pentanol, n-5-hydroxypentyl carbamic acid tert-butyl ester, tert-butyl 5-hydroxypentylcarbamate, carbamic acid, 5-hydroxypentyl-, 1,1-dimethylethyl ester, n-boc-5-aminopentanol, 5-boc-amino 1-pentanol, acmc-209ox9
PubChem CID 4456136
IUPAC Name tert-butyl N-(5-hydroxypentyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCO

Specifications

Color Colorless
Quantity 25 g
Formula Weight 203.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-(tert-Butoxycarbonylamino)-1-pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.