Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich 5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
1-(4-Chlorophenyl)ethylamine is a chiral amine that may be used in the preparation of N-phenylacetyl-(R)-1-(4-chlorophenyl)ethylamine via enantioselective acylation in aqueous medium and in the presence of penicillin acylase from Alcaligenes faecalis.
Specifications
Specifications
| CAS | 1135283-53-2 |
| Molecular Formula | C9H5F3N2O |
| Linear Formula | C9H5F3N2O |
| MDL Number | MFCD11841013 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 214.14 |
Product Title
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'