Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Trifluoromethyl-m-phenylenediamine, 97+%, Thermo Scientific™

Catalog No. AAB2366803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2366803 1 g
AAB2366806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2366803 Supplier Thermo Scientific Chemicals Supplier No. B2366803

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-53-6
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00014776
InChI Key KZSXRDLXTFEHJM-UHFFFAOYSA-N
Synonym 3,5-diaminobenzotrifluoride, 5-trifluoromethyl benzene-1,3-diamine, 5-trifluoromethyl-1,3-phenylenediamine, 5-trifluoromethyl-1,3-benzenediamine, 3,5-benzotrifluorodiamine, unii-z89rd529kk, 5-trifluoromethyl-benzene-1,3-diamine, 1,3-benzeneamine, 5-trifluoromethyl, 1,3-benzenediamine, 5-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-3,5-diamine
PubChem CID 67781
IUPAC Name 5-(trifluoromethyl)benzene-1,3-diamine
SMILES C1=C(C=C(C=C1N)N)C(F)(F)F

Specifications

Melting Point 86°C to 90°C
Quantity 1 g
UN Number UN2811
Beilstein 2966748
Formula Weight 176.14
Percent Purity ≥97%
Chemical Name or Material 5-Trifluoromethyl-m-phenylenediamine
Hazard Category H302+H332-H311-H315-H318-H335
Hazard Statement GHS H Statement
H311-H318-H302-H332-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-708-8
RTECSNumber CZ1606000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.