Learn More
Sigma Organic Chemistry 5-trifluoromethyl oxind | 100mg | 71293-62-4 | MFCD02179602
Supplier: Sigma Organic Chemistry CDS008597100MG

5-trifluoromethyl oxind , 100mg
About this Item:
CAS #: 71293-62-4
MDL #: MFCD02179602
Molecular Weight: 201.15
UNSPSC Code: 12352200
Chemical Formula: C9H6F3NO
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.