Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Trifluoromethyl)uracil 97.0+%, TCI America™
SDP

Catalog No. T16221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16221G 1 g
T16225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16221G Supplier TCI America Supplier No. T16221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54-20-6
Molecular Formula C5H3F3N2O2
Molecular Weight (g/mol) 180.09
MDL Number MFCD00006024
InChI Key LMNPKIOZMGYQIU-UHFFFAOYSA-N
Synonym 5-trifluoromethyl uracil, 5-trifluoromethyluracil, 5-trifluoromethyl pyrimidine-2,4 1h,3h-dione, trifluoromethyluracil, 2,4 1h,3h-pyrimidinedione, 5-trifluoromethyl, 5-tfmu, uracil, 5-trifluoromethyl, 5-trifluoromethyl pyrimidine-2,4-diol, 5-trifluoromethyl-uracil
PubChem CID 5899
IUPAC Name 5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES FC(F)(F)C1=CNC(=O)NC1=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 180.09
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Trifluoromethyl)uracil
RTECSNumber YR1750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.