Learn More
Ist Engineering 5 UM FILTER TUBE
Supplier:  Ist Engineering 538850
Encompass_Preferred
5um DNA filter cartridge with 2mL microcentrifuge collection tube. Easily removes particulate matter such as gel debris from sample volume. Maximum G-Force: 10,000xG. Bag of 50.
Catalog No. NC0406588
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.