Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Vinyl-2-norbornene, 98%, stabilized

Catalog No. AC164860250
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164860250 25 mL Glass bottle
AC164865000 500 mL Glass bottle
AC164860025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164860250 Supplier Thermo Scientific Chemicals Supplier No. 164860250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 3048-64-4 | C9H12 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3048-64-4
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00080691
InChI Key INYHZQLKOKTDAI-UHFFFAOYSA-N
Synonym 5-vinyl-2-norbornene, 2-vinylnorbornene, vinylnorbornene, 5-vinylnorbornene, 2-vinyl-5-norbornene, 2-norbornene, 5-vinyl, 5-vinylnorborn-2-ene, bicyclo 2.2.1 hept-2-ene, 5-ethenyl, vinylnorbornene van, 5-vinylbicyclo 2.2.1 hept-2-ene
PubChem CID 18273
IUPAC Name 5-ethenylbicyclo[2.2.1]hept-2-ene
SMILES C=CC1CC2CC1C=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -80°C
Boiling Point 141°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.482
Quantity 25 mL
Stabilizer 100 to 200ppm t-butylcatechol
Formula Weight 120.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Vinyl-2-norbornene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 221-259-8
RTECSNumber RC0350000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.