Learn More
Exel International 50 to 60cc Syringes
Catalog No. 1484137
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Connection Type:
Catheter
Leur Slip
Luer Lock
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Connection Type
14-841-37 Sterile Leur Slip
14-841-68 Non-sterile Catheter
14-841-36 Sterile Luer Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-841-37 Supplier Exel International Supplier No. 26301
Add to Cart
Add to Cart
This item is not returnable. View return policy
Latex free;Sterile;Non-toxic;Pyrogen free;Positive plunger stop to prevent spillage when drawing;Smooth plunger motion;Polypropylene construction.
Specifications
Certifications/Compliance Pyrogen Free
Latex Free Yes
Product Type Syringe
Volume (Metric) 50 to 60 mL
Connection Type Leur Slip
Sterility Sterile