Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific 500 mL Wide-Mouth HDPE Sample Bottles
SDP

Catalog No. 18366484 Shop All New Pig Corporation Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
18-366-484 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-484 Supplier United Scientific Supplier No. BTL102

missing translation for 'validMainframeMsg'

Reusable polypropylene bottles for collecting, storing or shipping stormwater samples, reagents, and other liquids.

  • Wide-mouth design makes filling these polypropylene bottles convenient
  • Polypropylene linerless cap
  • Leakproof
  • Easy to fill
  • Autoclavable
  • Translucent for easy viewing of liquid levels
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals
  • Holds 500 mL per bottle
  • Includes 12 bottles

Specifications

Capacity (Metric) 500 mL
Color Translucent
Material Polypropylene
No. per Case 12/Cs.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Fit for Purpose Certified Quality
Closure Material Polypropylene
Closure Size 1.75 in.
Closure Type Cap
Autoclavable Autoclavable
Shape Straight-sided
Disposable Reusable
Product Type Water Sample Collection Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.