Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 5000 Series 0.03μm to 0.30μm Polymer Particle Suspensions

Catalog No. 09980256
Change view
Click to view available options
Diameter (Metric):
0.03 μm
0.06 μm
0.08 μm
0.09 μm
0.10 μm
0.11 μm
0.12 μm
0.14 μm
0.16 μm
0.17 μm
0.20 μm
0.22 μm
0.24 μm
0.26 μm
0.3 μm
Quantity:
15 mL
100 mL
1000 mL
40 product options available for selection
Product selection table with 40 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Quantity
09-980-256 0.11 μm 1000 mL
09-980-239 0.03 μm 15 mL
09-980-240 0.03 μm 100 mL
09-980-241 0.03 μm 1000 mL
09-980-242 0.06 μm 15 mL
09-980-243 0.06 μm 100 mL
09-980-244 0.06 μm 1000 mL
09-980-245 0.08 μm 15 mL
09-980-248 0.09 μm 15 mL
09-980-249 0.09 μm 100 mL
09-980-250 0.09 μm 1000 mL
09-980-251 0.10 μm 15 mL
09-980-252 0.10 μm 100 mL
09-980-253 0.10 μm 1000 mL
09-980-254 0.11 μm 15 mL
09-980-255 0.11 μm 100 mL
09-980-257 0.12 μm 15 mL
09-980-258 0.12 μm 100 mL
09-980-259 0.12 μm 1000 mL
09-980-260 0.14 μm 15 mL
09-980-261 0.14 μm 100 mL
09-980-262 0.14 μm 1000 mL
09-980-263 0.16 μm 15 mL
09-980-264 0.16 μm 100 mL
09-980-265 0.16 μm 1000 mL
09-980-266 0.17 μm 15 mL
09-980-267 0.17 μm 100 mL
09-980-268 0.17 μm 1000 mL
09-980-269 0.20 μm 15 mL
09-980-270 0.20 μm 100 mL
09-980-271 0.20 μm 1000 mL
09-980-272 0.22 μm 15 mL
09-980-273 0.22 μm 100 mL
09-980-274 0.22 μm 1000 mL
09-980-275 0.24 μm 15 mL
09-980-278 0.26 μm 15 mL
09-980-279 0.26 μm 100 mL
09-980-280 0.26 μm 1000 mL
09-980-281 0.3 μm 15 mL
09-980-282 0.3 μm 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 40 options available.
40 options
Catalog No. 09-980-256 Supplier Thermo Scientific™ Supplier No. 5011C
Add to Cart
Add to Cart

Optimize research in light scattering, microporous filter checking, and aerosol particle generation with these particle suspensions.

Optimize research in light scattering, microporous filter checking, and aerosol particle generation with Thermo Scientific™ 5000 Series 0.03μm to 0.30μm Polymer Particle Suspensions. These particles are designed to meet the need for particulate materials with a variety of particle sizes and properties.

The polystyrene microspheres have a density of 1.05g/cm3 and a refractive index of 1.59 at 589nm. Particle diameters are measured by optical microscopy, photon correlation spectroscopy or light scattering. They are packaged as aqueous suspensions at 10% solids by weight. The 5000 Series particles range up to 3.1 microns in size. If you require larger particles, please refer to our 7000 Series Copolymer Microsphere Suspensions.

Ordering Information:

15mL, 100mL, and 1000mL bottle sizes available. 15mL (“A” Part No. bottles), 100mL (B bottles) and 1000mL (C bottles) are packaged to order; example (Part No. 5003B and 5003C). Please allow 10 working days when ordering these fill volumes.

Includes:

Material Safety Data Sheet (MSDS) and general product handling insert sheet

Recommended for:

High concentration polymer particles < 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications.

TRUSTED_SUSTAINABILITY

Specifications

Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
Concentration 10%
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles useful for such applications as filter evaluation, checking and testing, light scattering research, fluid mechanics research, aerosol particle generation, dispersion studies, and many other research and development projects. They are not intended for use in instrument calibration.
Color White or Colorless
For Use With (Application) Aerosol Particle Generation, Dispersion, Filter Evaluation, Light Scattering, Particle Generation
Material Polystyrene
Product Type Polymer Particle Suspensions
Includes General product handling insert sheet
Packaging 1000 mL
Particle Density 1.05g/cm3
Quantity 1000 mL
Refractive Index 1.59 at 589nm (25°C)
Stability Product stable for a minimum of 24 months
Diameter (Metric) 0.11 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.