Learn More
Description
Includes
Organic vapor cartridge and reusable storage bag.
- Ideally suited for intermittent use, plant shutdowns, and applications where respirators wear out or become difficult to clean in short periods of time
- Color coded for easy size identification
- Can be used with NIOSH-approved 3M filters N95, P95, and P100 for oil and non-oil particles
- For convenience and simplicity, some respirator assemblies are packaged with 3M Filters P95 or P100 along with 3M Filter Retainer 501 or Filter Adapter 502
- Some styles feature acid gas and/or organic vapor protection
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Color | Black |
| Material | Thermoplastic Elastomer |
| Product Type | Half-Mask Respirator |
| Size | Large |
| Facepiece Material | Thermoplastic Elastomer |
| For Use With (Application) | Spray painting, pesticides, agriculture, construction and transportation |
| For Use With (Equipment) | 3M filters N95, P95, and P100 for oil and non-oil particles |
| Includes | Organic vapor cartridge, reusable storage bag |
| Model | 53P71 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.