Learn More
Apexbio Technology LLC WP1066 857064-38-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A41405.1
Encompass_Preferred
WP1066 (CAS 857064-38-1) is a small molecule analog of AG490 and functions primarily as an inhibitor of Janus kinase 2 (JAK2) It disrupts JAK2 signaling by reducing phosphorylation and promoting degradation of the JAK2 protein thereby inhibiting downstream STAT3/STAT5 transcription factors and the phosphoinositide-3 kinase pathway WP1066 has demonstrated antiproliferative and pro-apoptotic activity in models of acute myeloid leukemia (AML) including patient-derived AML cells and AML cell lines (OCIM2 K562) Additionally WP1066 shows inhibitory effects against tumor growth and angiogenesis in renal cell carcinoma (RCC) model systems highlighting its potential for hematologic cancers and RCC therapeutic research
Catalog No. 50-000-61424
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.