Learn More
Apexbio Technology LLC (6-)ε-Aminocaproic acid 60-32-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B20765.1
Encompass_Preferred
6-epsilon-Aminocaproic acid is a small-molecule inhibitor targeting plasminogen activators It is designed to competitively inhibit the conversion of plasminogen to plasmin thereby regulating the fibrinolytic pathway and hemostatic processes 6-epsilon-Aminocaproic acid exerts its biological activity primarily through antifibrinolytic inhibition by blocking plasmin-mediated fibrin degradation Based on these pharmacological properties 6-epsilon-Aminocaproic acid holds research potential in coagulation studies thrombosis bleeding disorders and the investigation of pathological fibrinolysis mechanisms
Catalog No. 50-000-89391
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.