Learn More
Apexbio Technology LLC Nedaplatin 95734-82-0 25mg
SDP
Supplier:  Apexbio Technology LLC B198625
Encompass_Preferred
Nedaplatin (CAS 95734-82-0) is a platinum-based compound structurally related to cisplatin developed as a DNA-damaging agent It exerts its activity by forming platinum-DNA adducts leading to DNA crosslinking and disruption of DNA replication and transcription processes in tumor cells In cellular assays nedaplatin has demonstrated inhibition of tumor colony formation with an IC50 of 94 M This compound is utilized in biomedical research to investigate mechanisms of DNA damage response and to evaluate anticancer activities of platinum-based chemotherapeutics
Catalog No. 50-000-93286
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.