Learn More
Apexbio Technology LLC Alantolactone 546-43-0 20mg
Supplier: Apexbio Technology LLC N266020
Alantolactone (CAS 546-43-0) is a sesquiterpene lactone primarily isolated from the roots of Inula helenium L and leaves of Inula grandis This compound is known to modulate multiple cellular pathways including the inhibition of NF- B signaling and the induction of apoptosis via reactive oxygen species (ROS) generation Alantolactone has demonstrated anti-inflammatory and antiproliferative activities in various in vitro models making it a valuable tool for investigating molecular mechanisms underlying inflammation and cancer The compound should be stored in a sealed container under cool and dry conditions for optimal stability
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.