Learn More
Apexbio Technology LLC HyperFluor™ 555 Goat Anti-Rabbit IgG (H+L) Antibody 100ul
SDP
Supplier:  Apexbio Technology LLC K33090.1
Encompass_Preferred
HyperFluor 555 Goat Anti-Rabbit IgG HplusL Antibody is an affinity-purified polyclonal antibody targeting rabbit immunoglobulin G (IgG) heavy and light chains It is designed to recognize and bind to primary antibodies raised in rabbits thereby enabling indirect detection and signal amplification HyperFluor 555 Goat Anti-Rabbit IgG HplusL Antibody exerts its biological activity primarily through specific binding to rabbit IgG (H L) and is conjugated with the HyperFluor 555 fluorescent dye (excitation/emission 555/568 nm) Based on these properties HyperFluor 555 Goat Anti-Rabbit IgG HplusL Antibody holds research potential in fluorescence microscopy immunofluorescence staining flow cytometry and related fluorescence-based assays
Catalog No. 50-000-97148
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.