Learn More
Medchemexpress LLC Clozapine-d8 | 1185053-50-2 | 99.7% | C18H11D8ClN4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14539S10MG
Encompass_Preferred
Clozapine-d8 is the deuterium-labeled isotopologue of clozapine supplied as an analytical reference standard for pharmaceutical research. It is intended for use as an internal standard in quantitative LC-MS assays, method development, and pharmacokinetic studies where isotopic labeling improves accuracy, precision, and traceability of measurements.
  • High isotopic labeling to support accurate internal standardization.
  • Intended for LC-MS method development and quantitative assays.
  • Useful for pharmacokinetic and ADME studies requiring isotopic tracers.
  • Provided in small-mass formats suitable for analytical workflows.
  • Typically supplied with analytical documentation such as COA, HNMR, and LCMS.
Catalog No. 50-002-59574
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.