Learn More
Medchemexpress LLC E/Z-FK-506 26,28-allylic ester rearrangement impurity | 131944-48-4 | >90.0% | 804.02 | C44H69NO12 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW58588610MG
Encompass_Preferred
(E/Z)-FK-506 26,28-allylic ester rearrangement impurity (FK506-A) is an impurity of tacrolimus used as a reference standard for analytical and stability studies. It is intended for research and analytical applications to identify, quantify, and qualify impurity profiles; consult the certificate of analysis for storage and handling details.
  • Reference impurity of tacrolimus for analytical method development.
  • Suitable for impurity identification, reporting, and qualification studies.
  • High-purity material for reliable chromatographic and mass spectrometric analysis (>90.0%).
  • Supplied as a solid for laboratory use; consult the certificate of analysis for storage instructions.
  • Molecular formula C44H69NO12 and molecular weight 804.02 for compound identification.
Catalog No. 50-002-84640