Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 3',5'-diphosphate | 2922-95-4 | 99.7% | 404.16 g/mol | C9H14N2O12P2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY15482810MG

Encompass_Preferred

Uridine 3',5'-diphosphate is a nucleotide derivative used as a competitive RNase inhibitor in biochemical research. It is supplied as a powdered reagent for laboratory use and is intended for research purposes only. Key chemical identifiers include CAS 2922-95-4, molecular formula C9H14N2O12P2, and molecular weight 404.16 g/mol.

  • High purity (99.65%).
  • Powder form suitable for long-term storage.
  • Stable at -20°C for extended storage; in solvent stability guidelines available.
  • Functionally used as a competitive RNase inhibitor in biochemical assays.
  • Well-characterized molecular identity for reproducible research results.
  • Intended for research use only; not for clinical or human use.

Catalog No. 50-002-85863


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.