Learn More
Medchemexpress LLC 5,6-O-isopropylidene-L-ascorbic acid | 15042-01-0 | MFCD00010552 | 99.0% | 216.19 g/mol | C9H12O6 | 1 ML
Supplier: Medchemexpress LLC HYW01414110MM/1ML
5,6-O-Isopropylidene-L-ascorbic acid is a protected derivative of L-ascorbic acid used in research applications. The compound provides greater chemical stability than ascorbic acid and is supplied as a ready-to-use 10 mM solution in DMSO or as a solid for reconstitution, making it suitable for biochemical and molecular biology studies.
- Protected ascorbic acid derivative with antioxidant properties.
- Improved stability compared with unprotected ascorbic acid.
- Available as 10 mM solution in DMSO or as solid for preparation.
- High purity suitable for biochemical and molecular biology studies.
- Molecular formula C9H12O6; molecular weight 216.19 g/mol.
- Supplied in lab-scale pack sizes for convenient research use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.