Learn More
Fluotics Agilent Tapestation 2200 Compatible P30, filter/sterile
Supplier: Fluotics TPS420030.F
Agilent Tapestation Compatible P30 filter/sterile For Tapestation 4150/4200 Working Volume 15uL Tip Color Clear Tip Box Compare To 5067-5599 50 racks/case with 112 tips per rack