Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Proshield™ Basic Lab Coats

Catalog No. 19162605 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-605 7X-Large
19-162-604 6X-Large
19-162-603 5X-Large
19-162-600 2X-Large
19-162-601 3X-Large
19-162-602 4X-Large
19-162-606 Large
19-162-609 X-Large
19-162-608 Small
19-162-607 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-162-605 Supplier DuPont™ Supplier No. PB212SBU7X003000
Add to Cart
Add to Cart
This item is not returnable. View return policy

Disposable labcoats are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications.

  • Made of spunbond meltblown spunbond (SMS)
  • Optimized for comfort, softness and breathability
  • For general maintenance, janitorial, and cleaning

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size 7X-Large
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Resistance Light Liquid Splash, Particulates
Style Standard Length
No. of Pockets 2
Collar Type Traditional
Model ProShield 10
For Use With (Application) General Maintenance, Janitorial, Cleaning
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.