Learn More
Sino Biological Mouse CSF1R / MCSF Receptor / CD115 Protein (His Tag), Biotinylated 20ug
Supplier:  Sino Biological 50059M08HB20
Encompass_Preferred
A DNA sequence encoding the mouse CSF1R (NP_001032948.2) extracellular domain (Met1-Ser511) was fused with a polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vitro.
Catalog No. 50-162-1255
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.