Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes - Nitrogen Dioxide

Catalog No. 17985270 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
2 to 100ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range
17-985-270 2 to 100ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-985-270 Supplier Dräger™ Supplier No. 6719101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Short-term detector tube for the manual or automatic measurement of nitrogen dioxide gas concentrations.

Dräger Tubes are glass vials filled with a chemical reagent that reacts to a specific chemical for gas detection. These short-term detector tubes are used manually with the Dräger Accuro gas detection pump or automatically with the Accuro 2000 or Quantimeter 1000.

  • Gas Measured: Nitrogen dioxide
  • Standard Measurement Range: 5 to 100 ppm
  • Number of Strokes (n): 5
  • Measurement Time: Approximately 1 min
  • Standard Deviation: ±10 to 15%
  • Color Change: Yellow-green to blue-gray
  • Temperature Range: 10°C to 40°C
  • Humidity: Max. 30 mg H2O/L
  • Reaction Principle: NO2 + Diphenylbenzidine → blue-gray reaction product
  • Cross Sensitivity: Ozone or chlorine do not interfere within the range of their occupational exposure limits. Higher concentrations are indicated with different sensitivity. Nitrogen monoxide is not indicated.

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Nitrogen Dioxide 2/c
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range 30mg/L
Measurement Range 2 to 100ppm
No. of Strokes 5/10
Quantity 10
Response Time 1 to 2 min.
Temperature Range (English) 50°F to 104°F
Temperature Range (Metric) 10° to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.