Learn More
Ace Glass, Inc. 500mL filter flask, #25 Ace-Thred top, Ace-Safe hose connection
SDP
Supplier:  Ace Glass, Inc. 370008
Encompass_Preferred
500mL filter flask with #25 Ace-Thred top and an Ace-Safe hose connection. For use with the ACE borosilicate glass filtration apparatus that accomm ODates 25mm membrane filters.
Catalog No. 50-978-186
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.