Learn More
Alkali Scientific CellPro™ Vacuum Filtration Flasks, 500mL, 0.22µm Membrane, Individually Wrapped, 12/cs
SDP
Supplier:  Alkali Scientific V50022
Encompass_Preferred
CellPro™ Filtration Flasks are available in filter medium PES 0.22µm. The screw-on funnel has a lid, the filtration section has a graduated connection that fit multiple hoses, and the receiving bottle has indentations for easy gripping and a separate cap. Both the funnel and receiver feature engraved graduation marks to ensure accuracy, and are gamma sterilized, pyrogen free.
Catalog No. NC1934038
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.