Learn More
NEST SCIENTIFIC USA 500ML TUBE/BI-TRANSFER CAP
Supplier:  NEST SCIENTIFIC USA C61022EAB050A
Encompass_Preferred
NEST launches a new version of liquid transfer system to provide an aseptic transfer solution for pharmaceutical, biotech and laboratories, enabling closed injection of liquid. Standard Luer head is used in the joint, making the operation easier and more convenient. TPE material of tubes is in line with the requirements for industrial pharmaceutical enterprises, and has passed the dissolution test carried out by a certified third party according to the complete BPOG dissolution test criteria.Product Features1. Complete varieties comprise full specifications.2. The transfer cap comes out of integrated injection molding, whose congruence guarantees its adaptation to other brands.3. PETG material of the wall is of high transparency, strong mechanical strength, resistant to low temperature and convenient for observation and transportation4. The unique thick collar design of bottle neck makes inside operation easier.5. DNase/RNase/Pyrogen free.Material: PETG Bottle & HDPE Cap
Catalog No. NC2887715