Learn More
Sino Biological Mouse Galectin-1 / LGALS1 Protein 1mg
Supplier:  Sino Biological 50100MNAE1
Encompass_Preferred
A DNA sequence encoding the mouse Lgals1 (P16045) (Met 1-Glu 135) was expressed and purified.
Catalog No. 50-162-1344
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.