Learn More
Sino Biological Mouse IL-1 beta / IL1B Protein 100ug
Supplier:  Sino Biological 50101MNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of mouse IL1? (P10749) (Val 118-Ser 269) was expressed, with an initial Met at the C-terminus.
Catalog No. 50-162-1350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.