Learn More
ACROBiosystems Recombinant Protein;Human Latent TGF-beta 1 / TGFB1 Protein;HEK293;50UG;TG1,Latent TGFB1,TGFB1,TGF B1,TGF-B1,TGF-beta1,TGF beta1,TGFbeta1,CED,DPD1,DPD-1,LAP,TGFB,TGFbeta,TGF-β1,TGFβ1,TGF-beta,TGF beta,Latent TGF-beta 1
Supplier:  ACROBiosystems TG1H524X50UG
Encompass_Preferred
Recombinant Protein;Human Latent TGF-beta 1 / TGFB1 Protein;50UG;Human TGFB1, His Tag (TG1-H524x) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P01137-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 42.6 kDa (monomer). The protein migrates as 37-45 kDa (LAP) and 13 kDa (TGFB1) under reducing (R) condition (SDS-PAGE) due to glycosylation and Interchain disulfide bond.;Immobilized Human TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB6, His Tag, Avi Tag (Cat. No. IT6-H82E4) with a linear range of 1-20 ng/mL (QC tested).;Immobilized Human TGFB1, His Tag (Cat. No. TG1-H524x) at 5 μg/mL (100 μL/well) can bind Human LRRC32, Fc Tag (Cat. No. LR2-H5256) with a linear range of 39-625 ng/mL (Routinely tested).;Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) inhibits the IL-4-dependent proliferation of TF-1 cells. The ED50 for this effect is 0.293-0.406
Catalog No. 50-113-8264
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.