Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human BAFF / TNFSF13B / CD257 Protein, Avi Tag (Avitag™);HEK293;25UG;BAF,TNFSF13B,BAFF,BLYS,CD257,DTL,TALL-1,TALL1,THANK,TNFSF20,ZTNF4,TNFSF-13B,ZTNF-4
Supplier:  ACROBiosystems BAFH82Q225UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human BAFF / TNFSF13B / CD257 Protein, Avi Tag (Avitag™);25UG;Biotinylated Human BAFF, His Tag (BAF-H82Q2) is expressed from human 293 cells (HEK293). It contains AA Ala 134 - Leu 285 (Accession # AAH20674.1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 29.2 kDa. The protein migrates as 27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 2 μg/mL (100 μL/well) can bind Biotinylated Human BAFF (Cat. No. BAF-H82Q2) with a linear range of 0.24-3.9 ng/mL (QC tested).;Immobilized Human BCMA, His Tag (Cat. No. BCA-H522y) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BAFF (Cat. No. BAF-H82Q2) with a linear range of 0.24-3.9 ng/mL (Routinely tested).
Catalog No. 50-113-8444
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.