Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD30 / TNFRSF8 Protein, Avi Tag (Avitag™);HEK293;25UG;CD0,TNFRSF8,CD30,TNFRSF-8,D1S166E,Ki-1
Supplier: ACROBiosystems CD0H82E625UG
Recombinant Protein;Biotinylated Human CD30 / TNFRSF8 Protein, Avi Tag (Avitag™);25UG;Biotinylated Human CD30, His Tag (CD0-H82E6) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 41.1 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD30 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD30, His Tag (Cat. No. CD0-H82E6) with a linear range of 0.4-6.5 ng/mL (Routinely tested).;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD30, His Tag (Cat. No. CD0-H82E6) with a linear range of 10-78 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.