Learn More
Thermo Scientific 6-Port Valve Assembly Kit [6 EA]
Supplier: Thermo Scientific 075974
6-port valve assembly kit for gas chromatography enables precise control over sample injection and column switching with ports allowing connection to multiple analytical columns or sample injection systems ™